CO1H324A6644 CombiPhos Catalysts Inc

2-Bromo-6-(2-pyridyl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223034-97-5
  • Molecular Weight : 361.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart