CO1H324A664D CombiPhos Catalysts Inc

2-Bromo-6-(thiazol-5-yl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223046-79-3
  • Molecular Weight : 367.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart