CO1H324A664E CombiPhos Catalysts Inc

2-(n-Propyl)-6-trifluoromethylpyridine-4-boronic acid pinacol ester

  • Cas Number : 2223031-14-7
  • Molecular Weight : 315.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart