CO1H324A6654 CombiPhos Catalysts Inc

4-Methyl-5-ethylpyridine-3-boronic acid pinacol ester

  • Cas Number : 2223041-56-1
  • Molecular Weight : 247.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart