CO1H324A665A CombiPhos Catalysts Inc

2-(sec-Butyl)-6-cyanopyridine-4-boronic acid pinacol ester

  • Cas Number : 2223009-41-2
  • Molecular Weight : 286.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart