CO1H324A665C CombiPhos Catalysts Inc

2-Chloro-6-(n-propyl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223032-49-1
  • Molecular Weight : 281.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart