CO1H324A6664 CombiPhos Catalysts Inc

2-Trifluoromethyl-6-(n-propoxy)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223046-96-4
  • Molecular Weight : 331.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart