CO1H324A666B CombiPhos Catalysts Inc

3-Methyl-2-ethylpyridine-4-boronic acid pinacol ester

  • Cas Number : 2223041-83-4
  • Molecular Weight : 247.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart