CO1H324A6673 CombiPhos Catalysts Inc

4-Methyl-5-(tert-butyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223032-82-2
  • Molecular Weight : 275.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart