CO1H324A667A CombiPhos Catalysts Inc

4-Methyl-6-(tert-butyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223040-67-1
  • Molecular Weight : 275.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart