CO1H324A667D CombiPhos Catalysts Inc

2-Methyl-5-(n-propyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223044-91-3
  • Molecular Weight : 261.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart