CO1H324A6684 CombiPhos Catalysts Inc

5-Methyl-6-(iso-propyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223044-12-8
  • Molecular Weight : 261.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart