CO1H324A6686 CombiPhos Catalysts Inc

5-Methyl-6-(iso-butyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223048-54-0
  • Molecular Weight : 275.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart