CO1H324A6696 CombiPhos Catalysts Inc

5-(tert-Butyl)-6-methoxypyridine-3-boronic acid pinacol ester

  • Cas Number : 2223033-59-6
  • Molecular Weight : 291.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart