CO1H324A6698 CombiPhos Catalysts Inc

2-Amino-5-chloropyridine-4-boronic acid pinacol ester

  • Cas Number : 1222522-38-4
  • Molecular Weight : 254.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart