CO1H324A669C CombiPhos Catalysts Inc

6-chloroimidazo[1,2-b]pyridazine-3-boronic acid pinacol ester

  • Cas Number : 2223035-24-1
  • Molecular Weight : 279.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart