CO1H324A669D CombiPhos Catalysts Inc

2-(1H-Tetraazol-5-yl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223044-96-8
  • Molecular Weight : 273.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart