CO1H324A66A2 CombiPhos Catalysts Inc

1-(6-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)ethan-1-one

  • Cas Number : 2223040-73-9
  • Molecular Weight : 261.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart