CO1H324A66A4 CombiPhos Catalysts Inc

2,6-Di(acetyl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223044-89-9
  • Molecular Weight : 289.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart