CO1H324A66BB CombiPhos Catalysts Inc

tert-butyl 3-(6-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)pyrrolidine-1-carboxylate

  • Cas Number : 2222995-87-9
  • Molecular Weight : 408.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart