CO1H324A66C7 CombiPhos Catalysts Inc

4-(N-Boc-Pyrrolidin-3-yl)phenylboronic acid pinacol ester

  • Cas Number : 1802370-70-2
  • Molecular Weight : 373.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart