CO1H324A66DF CombiPhos Catalysts Inc

4-(Oxolan-2-yl)phenylboronic acid pinacol ester

  • Cas Number : 1857371-54-0
  • Molecular Weight : 274.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart