CO1H324A66E4 CombiPhos Catalysts Inc

2-(tetrahydrofuran-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-6-(trifluoromethyl)pyridine

  • Cas Number : 2223009-65-0
  • Molecular Weight : 343.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart