CO1H324A66E7 CombiPhos Catalysts Inc

2-Bromo-6-(Oxolan-2-yl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223003-38-9
  • Molecular Weight : 354.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart