CO1H324A66EC CombiPhos Catalysts Inc

methyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-6-(trifluoromethyl)isonicotinate

  • Cas Number : 1622217-28-0
  • Molecular Weight : 331.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart