CO1H324A66EF CombiPhos Catalysts Inc

2-Ethoxythiazole-5-boronic acid pinacol ester

  • Cas Number : 2096338-64-4
  • Molecular Weight : 255.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart