CO1H324A66F9 CombiPhos Catalysts Inc

2-(Cyclohexyloxy)thiazole-5-boronic acid pinacol ester

  • Cas Number : 1352652-25-5
  • Molecular Weight : 309.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart