CO1H324A66FE CombiPhos Catalysts Inc

2-(tert-Butyl)thiazole-5-boronic acid pinacol ester

  • Cas Number : 1426089-85-1
  • Molecular Weight : 267.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart