CO1H324A6703 CombiPhos Catalysts Inc

2-((tetrahydrofuran-3-yl)oxy)-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • Cas Number : 1315351-11-1
  • Molecular Weight : 291.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart