CO1H324A6715 CombiPhos Catalysts Inc

4,4,5,5-tetramethyl-2-(5-(tetrahydro-2H-pyran-4-yl)furan-2-yl)-1,3,2-dioxaborolane

  • Cas Number : 2223033-06-3
  • Molecular Weight : 278.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart