CO1H324A6718 CombiPhos Catalysts Inc

2-(5-isobutylfuran-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • Cas Number : 2223012-02-8
  • Molecular Weight : 250.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart