CO1H324A671A CombiPhos Catalysts Inc

3-(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)furan-2-yl)pyridine

  • Cas Number : 2223051-82-7
  • Molecular Weight : 271.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart