CO1H324A6723 CombiPhos Catalysts Inc

5-(3-Tolyl)furan-2-boronic acid pinacol ester

  • Cas Number : 2223040-86-4
  • Molecular Weight : 284.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart