CO1H324A672F CombiPhos Catalysts Inc

5-(6-Cyano-2-pyridyl)furan-2-boronic acid pinacol ester

  • Cas Number : 2223042-42-8
  • Molecular Weight : 296.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart