CO1H324A6730 CombiPhos Catalysts Inc

5-(6-Chloro-2-pyridyl)furan-2-boronic acid pinacol ester

  • Cas Number : 2223035-35-4
  • Molecular Weight : 305.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart