CO1H324A6735 CombiPhos Catalysts Inc

5-(3-Pyridyl)thiazole-4-boronic acid pinacol ester

  • Cas Number : 2223033-08-5
  • Molecular Weight : 288.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart