CO1H324A673A CombiPhos Catalysts Inc

2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(m-tolyl)pyrazine

  • Cas Number : 1402172-35-3
  • Molecular Weight : 296.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart