CO1H324A674C CombiPhos Catalysts Inc

5-(4-Methoxyphenyl)pyrazine-2-boronic acid pinacol ester

  • Cas Number : 1402227-15-9
  • Molecular Weight : 312.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart