CO1H324A675D CombiPhos Catalysts Inc

2-Ethyl-5-chloropyridine-3-boronic acid pinacol ester

  • Cas Number : 2223048-87-9
  • Molecular Weight : 267.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart