CO1H324A675E CombiPhos Catalysts Inc

2-(iso-Propyl)-5-chloropyridine-3-boronic acid pinacol ester

  • Cas Number : 2223047-49-0
  • Molecular Weight : 281.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart