CO1H324A6769 CombiPhos Catalysts Inc

2-Ethyl-5-trifluoromethylpyridine-3-boronic acid pinacol ester

  • Cas Number : 2223047-56-9
  • Molecular Weight : 301.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart