CO1H324A6776 CombiPhos Catalysts Inc

2-(tert-Butyl)-5-cyclopropylpyridine-3-boronic acid pinacol ester

  • Cas Number : 2223047-81-0
  • Molecular Weight : 301.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart