CO1H324A677A CombiPhos Catalysts Inc

2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(2-(trifluoromethyl)phenyl)pyrazine

  • Cas Number : 1402233-84-4
  • Molecular Weight : 350.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart