CO1H324A6785 CombiPhos Catalysts Inc

2-Hydroxy-5-fluorolpyridine-4-boronic acid pinacol ester

  • Cas Number : 2223048-48-2
  • Molecular Weight : 239.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart