CO1H324A6786 CombiPhos Catalysts Inc

2-Bromo-5-fluorolpyridine-4-boronic acid pinacol ester

  • Cas Number : 2223033-31-4
  • Molecular Weight : 301.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart