CO1H324A6798 CombiPhos Catalysts Inc

5-Ethyl-2-cyanopyridine-4-boronic acid pinacol ester

  • Cas Number : 2223049-00-9
  • Molecular Weight : 258.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart