CO1H324A6799 CombiPhos Catalysts Inc

5-(iso-Propyl-2-cyanopyridine-4-boronic acid pinacol ester

  • Cas Number : 2223045-30-3
  • Molecular Weight : 272.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart