CO1H324A679D CombiPhos Catalysts Inc

5-Bromo-2-(methoxycarbonyl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223041-16-3
  • Molecular Weight : 342

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart