CO1H324A67A9 CombiPhos Catalysts Inc

tert-butyl (5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazin-2-yl)carbamate

  • Cas Number : 1402227-80-8
  • Molecular Weight : 321.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart