CO1H324A67B7 CombiPhos Catalysts Inc

4-Fluoro-2-(tert-butoxy)pyridine-5-boronic acid pinacol ester

  • Cas Number : 2223045-25-6
  • Molecular Weight : 295.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart